4-(Allyloxy)benzophenone ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(4-(Allyloxy)phenyl)(phenyl)methanone, [4-(Allyloxy)phenyl](phenyl)methanone
A3242-1G
A3242-5G
TCIAA3242-1GEA
70
EUR
TCIAA3242-1G
TCIAA3242-5G
4-(Allyloxy)benzophenone ≥97.0% (by GC)
4-(Allyloxy)benzophenone
Formula:
C₁₆H₁₄O₂ Peso molecolare: 238,29 g/mol |
MDL Number:
MFCD07435384 CAS Number: 42403-77-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...