Stampa…

Adamantane ≥99.5% (by GC), purified by sublimation

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tricyclo[3.3.1.1(3,7)]decane

A3233-5G A3233-25G
TCIAA3233-5GEA 56 EUR
TCIAA3233-5G TCIAA3233-25G
Adamantane ≥99.5% (by GC), purified by sublimation
Adamantane
Formula: C₁₀H₁₆
Peso molecolare: 136,24 g/mol
MDL Number: MFCD00074719
CAS Number: 281-23-2
EINECS: 206-001-4
Merck Index: 14,00149

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR