Stampa…

1-Allyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Allyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide

A3210-25G A3210-5G
TCIAA3210-25GEA 242 EUR
TCIAA3210-25G TCIAA3210-5G
1-Allyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)
1-Allyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide
Formula: C₉H₁₁F₆N₃O₄S₂
Peso molecolare: 403,31 g/mol
MDL Number: MFCD11109343
CAS Number: 655249-87-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR