9-Aminoanthracene hydrochloride ≥98.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
Synonyms:
Anthracen-9-amine hydrochloride
A3208-1G
A3208-5G
TCIAA3208-1GEA
178
EUR
TCIAA3208-1G
TCIAA3208-5G
9-Aminoanthracene hydrochloride ≥98.0% (by total nitrogen basis)
9-Aminoanthracene hydrochloride
Formula:
C₁₄H₁₁N·HCl Peso molecolare: 229,71 g/mol |
MDL Number:
MFCD00050528 CAS Number: 209743-32-8 |
Risultati del test di specificazione
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...