Salmeterol xinafoate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Salmeterol 1-hydroxy-2-naphthoate
A3190-1G
A3190-250MG
TCIAA3190-1GEA
647
EUR
TCIAA3190-1G
TCIAA3190-250MG
Salmeterol xinafoate ≥98.0% (by HPLC, titration analysis)
Salmeterol xinafoate
Formula:
C₂₅H₃₇NO₄·C₁₁H₈O₃ Peso molecolare: 603,76 g/mol |
MDL Number:
MFCD00897708 CAS Number: 94749-08-3 Merck Index: 14,08337 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 % (total of Ion-Pair) |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...