Stampa…

Acenocoumarol ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Hydroxy-3-[1-(4-nitrophenyl)-3-oxobutyl]-2H-chromen-2-one

A3186-1G A3186-250MG A3186-50MG
TCIAA3186-1GEA 680 EUR
TCIAA3186-1G TCIAA3186-250MG TCIAA3186-50MG
Acenocoumarol ≥98.0% (by HPLC)
Acenocoumarol
Formula: C₁₉H₁₅NO₆
Peso molecolare: 353,33 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00137816
CAS Number: 152-72-7
EINECS: 205-807-3
Merck Index: 14,00029

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 3.50 to 4.50 %
Melting point 196.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR