Stampa…

Allyl (3-Methylbutoxy)acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (3-Methylbutoxy)acetic acid Allyl ester

A3185-5G A3185-25G
TCIAA3185-5GEA 131 EUR
TCIAA3185-5G TCIAA3185-25G
Allyl (3-Methylbutoxy)acetate ≥98.0% (by GC)
Allyl (3-Methylbutoxy)acetate
Formula: C₁₀H₁₈O₃
Peso molecolare: 186,25 g/mol
MDL Number: MFCD03424212
CAS Number: 67634-00-8
EINECS: 266-803-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c7f8ef4df65ec2f6a7be7c84c93933370dc9dd2&vid=5c7f8ef4df65ec2f6a7be7c84c93933370dc9dd2&prid=web&clid=SAPDAM

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR