2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₂₀H₂₁BO₂ |
MDL Number:
MFCD24039741 CAS Number: 709022-63-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...