Sakakin
Fornitore: TCI EUROPE
Synonyms:
(3R,4S,5S,6R)-2-(3-Hydroxy-5-methylphenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 3-Hydroxy-5-methylphenyl beta-D-Glucopyranoside, Orcinol glucoside
A3183-10MG
A3183-50MG
TCIAA3183-10MGEA
46
EUR
TCIAA3183-10MG
TCIAA3183-50MG
Sakakin
Sakakin
Formula:
C₁₃H₁₈O₇ Peso molecolare: 286,28 g/mol Punto di fusione: 132°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06794970 CAS Number: 21082-33-7 |
Risultati del test di specificazione
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...