Stampa…

3-Amino-4-methylphenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Amino-p-cresol, 5-Hydroxy-2-methylaniline

A3180-1G A3180-5G
TCIAA3180-1GEA 37 EUR
TCIAA3180-1G TCIAA3180-5G
3-Amino-4-methylphenol ≥98.0% (by GC)
3-Amino-4-methylphenol
Formula: C₇H₉NO
Peso molecolare: 123,16 g/mol
MDL Number: MFCD00075083
CAS Number: 2836-00-2
EINECS: 220-622-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=437ce6bb979904ce1570eb799766bdeab9616f96&vid=437ce6bb979904ce1570eb799766bdeab9616f96&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR