2-Amino-5-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-chloro-1,3-benzothiazol-2-amine
A3177-5G
A3177-1G
TCIAA3177-5GEA
132
EUR
TCIAA3177-5G
TCIAA3177-1G
2-Amino-5-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-chlorobenzothiazole
Formula:
C₇H₅ClN₂S Peso molecolare: 184,65 g/mol |
MDL Number:
MFCD00205355 CAS Number: 20358-00-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 199.0 to 203.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...