1-Allylpiperazine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
A3172-25G
A3172-5G
TCIAA3172-25GEA
254
EUR
TCIAA3172-25G
TCIAA3172-5G
1-Allylpiperazine ≥98.0% (by GC, titration analysis)
1-Allylpiperazine
Formula:
C₇H₁₄N₂ Peso molecolare: 126,2 g/mol Punto di ebollizione: 186°C Flash Pt: 51°C |
MDL Number:
MFCD00167970 CAS Number: 13961-36-9 UN: 2734 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...