4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A3164-25G
A3164-5G
TCIAA3164-25GEA
313
EUR
TCIAA3164-25G
TCIAA3164-5G
4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Amino-2-fluorobenzoic acid
Formula:
C₇H₆FNO₂ Peso molecolare: 155,13 g/mol |
MDL Number:
MFCD01569397 CAS Number: 446-31-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...