Stampa…

5-Bromo-3-fluoropyridin-2-amine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-5-bromo-3-fluoropyridine, 5-Bromo-3-fluoropyridin-2-amine

A3138-200MG A3138-1G
TCIAA3138-200MGEA 36 EUR
TCIAA3138-200MG TCIAA3138-1G
5-Bromo-3-fluoropyridin-2-amine ≥98.0% (by GC, titration analysis)
5-Bromo-3-fluoropyridin-2-amine
Formula: C₅H₄BrFN₂
Peso molecolare: 191,0011 g/mol
MDL Number: MFCD11848483
CAS Number: 748812-37-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR