Stampa…

3-(Acetylthio)propionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Acetylthio)propanoic acid

A3134-1G A3134-5G
TCIAA3134-1GEA 81 EUR
TCIAA3134-1G TCIAA3134-5G
3-(Acetylthio)propionic acid ≥98.0% (by GC, titration analysis)
3-(Acetylthio)propionic acid
Formula: C₅H₈O₃S
Peso molecolare: 148,18 g/mol
Punto di ebollizione: 128°C
MDL Number: MFCD06208418
CAS Number: 41345-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a946f2ccd3882232a79d201c1f8fb9c2852d1bca&vid=a946f2ccd3882232a79d201c1f8fb9c2852d1bca&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR