cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(2S,4S)-4-Azido-1-Boc-2-pyrrolidinecarboxylic acid, (2S,4S)-4-Azido-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid, cis-4-Azido-N-Boc-L-proline
A3129-50MG
A3129-200MG
TCIAA3129-50MGEA
39
EUR
TCIAA3129-50MG
TCIAA3129-200MG
cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline ≥98.0% (by HPLC, titration analysis)
cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline
Formula:
C₁₀H₁₆N₄O₄ Peso molecolare: 256,26 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD04115791 CAS Number: 132622-65-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Specific rotation [a]20/D | -23.0 to -27.0 deg (C=1, Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...