2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(2-(2-Aminoethoxy)ethoxy)acetic acid, H2N-PEG2-CH2COOH, 8-Amino-3,6-dioxaoctanoic acid, Amino-PEG2-acetic acid
A3120-250MG
A3120-1G
TCIAA3120-250MGEA
60
EUR
TCIAA3120-250MG
TCIAA3120-1G
2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)
2-[2-(2-Aminoethoxy)ethoxy]acetic acid
Formula:
C₆H₁₃NO₄ Peso molecolare: 163,17 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD13185897 CAS Number: 134978-97-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 124.0 to 128.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...