Stampa…

Almotriptan Malate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3-[2-(Dimethylamino)ethyl]-5-(pyrrolidin-1-ylsulphonylmethyl)-1H-indole Malate

A3111-100MG A3111-25MG
TCIAA3111-100MGEA 396 EUR
TCIAA3111-100MG TCIAA3111-25MG
Almotriptan Malate ≥98.0% (by HPLC)
Almotriptan Malate
Formula: C₁₇H₂₅N₃O₂S·C₄H₆O₅
Peso molecolare: 469,55 g/mol
MDL Number: MFCD08067740
CAS Number: 181183-52-8
Merck Index: 14,00304

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.50 to 9.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR