Stampa…

Acitretin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (2E,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid

A3097-50MG A3097-250MG
TCIAA3097-50MGEA 117 EUR
TCIAA3097-50MG TCIAA3097-250MG
Acitretin ≥98.0% (by HPLC, titration analysis)
Acitretin
Formula: C₂₁H₂₆O₃
Peso molecolare: 326,4 g/mol
Punto di fusione: 230°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00866632
CAS Number: 55079-83-9
EINECS: 259-474-4
UN: 3077
ADR: 9,III
Merck Index: 14,00112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=629f7c928a009a61be49d5603c8f7c670e8030ab&vid=629f7c928a009a61be49d5603c8f7c670e8030ab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR