Stampa…

5-Aminovaleric acid hydrobromide, low water content ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Homopiperidinic acid hydrobromide, 5-Aminopentanoic acid hydrobromide, 5-AVABr

A3094-1G A3094-5G
TCIAA3094-1GEA 81 EUR
TCIAA3094-1G TCIAA3094-5G
5-Aminovaleric acid hydrobromide, low water content ≥98.0% (by titrimetric analysis)
5-Aminovaleric acid hydrobromide
Formula: C₅H₁₁NO₂·HBr
Peso molecolare: 198,06 g/mol
CAS Number: 2173111-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e91d81ef5c7c506f8b84aecb0f967c9f6e483461&vid=e91d81ef5c7c506f8b84aecb0f967c9f6e483461&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 250 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR