Stampa…

5-Azoniaspiro[4.4]nonane chloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A3092-1G A3092-5G
TCIAA3092-1GEA 43 EUR
TCIAA3092-1G TCIAA3092-5G
5-Azoniaspiro[4.4]nonane chloride ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane chloride
Formula: C₈H₁₆ClN
Peso molecolare: 161,67 g/mol
Punto di fusione: 295°C
CAS Number: 98997-63-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=232fed7bb9974d647f6b517419bff27902c487b2&vid=232fed7bb9974d647f6b517419bff27902c487b2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR