Stampa…

2-Amino-5-pyrimidineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Aminopyrimidine-5-boronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A3087-1G A3087-5G
TCIAA3087-1GEA 61 EUR
TCIAA3087-1G TCIAA3087-5G
2-Amino-5-pyrimidineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Amino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₆BN₃O₂
Peso molecolare: 221,06 g/mol
MDL Number: MFCD06795685
CAS Number: 402960-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01c19d4eaf54cf0a65060d166e012c3b6b2e3fdd&vid=01c19d4eaf54cf0a65060d166e012c3b6b2e3fdd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR