5-Phenyl-1,3,4-oxadiazol-2-amine ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Amino-5-phenyl-1,3,4-oxadiazole
A3074-1G
A3074-5G
TCIAA3074-1GEA
38
EUR
TCIAA3074-1G
TCIAA3074-5G
5-Phenyl-1,3,4-oxadiazol-2-amine ≥97.0% (by GC, titration analysis)
5-Phenyl-1,3,4-oxadiazol-2-amine
Formula:
C₈H₇N₃O Peso molecolare: 161,16 g/mol |
MDL Number:
MFCD00126383 CAS Number: 1612-76-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...