Stampa…

2-Amino-5-chloro-3-iodopyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chloro-3-iodo-2-pyridinamine, 5-Chloro-3-iodo-2-pyridylamine, 5-Chloro-3-iodopyridin-2-amine

A3069-1G A3069-5G
TCIAA3069-1GEA 40 EUR
TCIAA3069-1G TCIAA3069-5G
2-Amino-5-chloro-3-iodopyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-chloro-3-iodopyridine
Formula: C₅H₄ClIN₂
Peso molecolare: 254,46 g/mol
MDL Number: MFCD07644578
CAS Number: 211308-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe1de862d2ed8c166f7e84b7cc887bd16194ff4c&vid=fe1de862d2ed8c166f7e84b7cc887bd16194ff4c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR