4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(4-Chlorophenyl)-5-methylthiazol-2-amine, 2-Amino-4-(4-chlorophenyl)-5-methylthiazole
A3066-1G
A3066-5G
TCIAA3066-1GEA
68
EUR
TCIAA3066-1G
TCIAA3066-5G
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine ≥98.0% (by GC)
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine
Formula:
C₁₀H₉ClN₂S Peso molecolare: 224,71 g/mol |
MDL Number:
MFCD00731832 CAS Number: 82632-77-7 |
Risultati del test di specificazione
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 145.0 to 148.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...