Stampa…

2-Amino-5-bromobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 5-Bromoanthranilaldehyde

A3063-200MG A3063-1G
TCIAA3063-200MGEA 32 EUR
TCIAA3063-200MG TCIAA3063-1G
2-Amino-5-bromobenzaldehyde ≥98.0% (by GC)
2-Amino-5-bromobenzaldehyde
Formula: C₇H₆BrNO
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD10696879
CAS Number: 29124-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c38df6caa86574ddc7e7be8264642f5c16a285e&vid=4c38df6caa86574ddc7e7be8264642f5c16a285e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR