Stampa…

2-Amino-3,5-dichlorobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

A3062-1G A3062-5G
TCIAA3062-1GEA 57 EUR
TCIAA3062-1G TCIAA3062-5G
2-Amino-3,5-dichlorobenzonitrile ≥98.0% (by GC)
2-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
Peso molecolare: 187,03 g/mol
MDL Number: MFCD00158947
CAS Number: 36764-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fc4886a3e7747d366654a0c394ba30966cd7ac2&vid=6fc4886a3e7747d366654a0c394ba30966cd7ac2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR