Stampa…

N-(5-Iodo-2-pyridinyl)acetamide ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Acetamido-5-iodopyridine, 2-Acetylamino-5-iodopyridine

A3050-1G A3050-5G
TCIAA3050-1GEA 103 EUR
TCIAA3050-1G TCIAA3050-5G
N-(5-Iodo-2-pyridinyl)acetamide ≥98.0% (by GC)
N-(5-Iodo-2-pyridinyl)acetamide
Formula: C₇H₇IN₂O
Peso molecolare: 262,05 g/mol
MDL Number: MFCD01315262
CAS Number: 66131-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb0ff6c4e967d9c02bde99fed060940467951460&vid=cb0ff6c4e967d9c02bde99fed060940467951460&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR