Stampa…

3-Amino-2,5-dibromopyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,5-Dibromopyridin-3-amine

A3048-1G A3048-5G
TCIAA3048-1GEA 36 EUR
TCIAA3048-1G TCIAA3048-5G
3-Amino-2,5-dibromopyridine ≥98.0% (by GC)
3-Amino-2,5-dibromopyridine
Formula: C₅H₄Br₂N₂
Peso molecolare: 251,9100 g/mol
MDL Number: MFCD09266224
CAS Number: 90902-84-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR