Stampa…

(4-Allyl-2-methoxyphenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A3037-1G A3037-5G
TCIAA3037-1GEA 67 EUR
TCIAA3037-1G TCIAA3037-5G
(4-Allyl-2-methoxyphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(4-Allyl-2-methoxyphenoxy)acetic acid
Formula: C₁₂H₁₄O₄
Peso molecolare: 222,24 g/mol
MDL Number: MFCD00017594
CAS Number: 6331-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a676f722f30f2e3fd655c7ad32dec907c847c3&vid=65a676f722f30f2e3fd655c7ad32dec907c847c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR