Stampa…

2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4'-Chloro-[1,1'-biphenyl]-2-amine

A3034-1G A3034-5G
TCIAA3034-1GEA 61 EUR
TCIAA3034-1G TCIAA3034-5G
2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)
2-Amino-4'-chlorobiphenyl
Formula: C₁₂H₁₀ClN
Peso molecolare: 203,67 g/mol
MDL Number: MFCD03424659
CAS Number: 1204-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb0cd51940068f81f9c76cbb7e7220f5ae1e0e04&vid=fb0cd51940068f81f9c76cbb7e7220f5ae1e0e04&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR