Stampa…

5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Chlorophenyl)-1-phenyl-1H-pyrazol-5-amine

A3033-1G A3033-5G
TCIAA3033-1GEA 58 EUR
TCIAA3033-1G TCIAA3033-5G
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole ≥98.0% (by HPLC, titration analysis)
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole
Formula: C₁₅H₁₂ClN₃
Peso molecolare: 269,73 g/mol
MDL Number: MFCD00197059
CAS Number: 19652-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ba31baecc96720aeb4461bffdee45f99548b454&vid=1ba31baecc96720aeb4461bffdee45f99548b454&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR