Stampa…

1-(2-Amino-4-methoxy-3-methylphenyl)ethanone ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2'-Amino-3'-methyl-4'-methoxyacetophenone

A3027-1G A3027-5G
TCIAA3027-1GEA 83 EUR
TCIAA3027-1G TCIAA3027-5G
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone ≥97.0% (by GC, titration analysis)
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone
Formula: C₁₀H₁₃NO₂
MDL Number: MFCD11042290
CAS Number: 912347-94-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR