Stampa…

5-Amino-3-bromo-2-methylpyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Amino-3-bromo-2-picoline

A2996-1G A2996-5G
TCIAA2996-1GEA 98 EUR
TCIAA2996-1G TCIAA2996-5G
5-Amino-3-bromo-2-methylpyridine ≥98.0% (by GC, titration analysis)
5-Amino-3-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
Peso molecolare: 187,0372 g/mol
MDL Number: MFCD08060462
CAS Number: 186593-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a946b431618dd6dd3cde6a27caf67ca8abbad1b&vid=6a946b431618dd6dd3cde6a27caf67ca8abbad1b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR