Stampa…

3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

A2993-200MG A2993-1G
TCIAA2993-200MGEA 57 EUR
TCIAA2993-200MG TCIAA2993-1G
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₉BN₂O₃
MDL Number: MFCD12923427
CAS Number: 893440-50-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR