Stampa…

2-Amino-6-methylpyrazine ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Methylpyrazin-2-amine

A2992-200MG A2992-1G
TCIAA2992-200MGEA 27 EUR
TCIAA2992-200MG TCIAA2992-1G
2-Amino-6-methylpyrazine ≥97.0% (by GC)
2-Amino-6-methylpyrazine
Formula: C₅H₇N₃
Peso molecolare: 109,13 g/mol
MDL Number: MFCD08235193
CAS Number: 5521-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26a9428d6ba3a03dfd625afb8884e500f6d06a33&vid=26a9428d6ba3a03dfd625afb8884e500f6d06a33&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR