3-Acetylpyridine N-Oxide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Acetylpyridine n-oxide
A2987-1G
A2987-5G
TCIAA2987-1GEA
79
EUR
TCIAA2987-1G
TCIAA2987-5G
3-Acetylpyridine N-Oxide ≥98.0% (by GC)
3-Acetylpyridine N-Oxide
Formula:
C₇H₇NO₂ Peso molecolare: 137,14 g/mol |
MDL Number:
MFCD00955798 CAS Number: 14188-94-4 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...