Stampa…

2-Amino-4-fluoropyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2976-1G A2976-5G
TCIAA2976-1GEA 113 EUR
TCIAA2976-1G TCIAA2976-5G
2-Amino-4-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-fluoropyridine
Formula: C₅H₅FN₂
Peso molecolare: 112,04 g/mol
MDL Number: MFCD09260906
CAS Number: 944401-77-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR