Stampa…

3-Amino-2-bromo-5-chloropyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromo-5-chloro-3-pyridylamine

A2973-5G A2973-25G
TCIAA2973-5GEA 43 EUR
TCIAA2973-5G TCIAA2973-25G
3-Amino-2-bromo-5-chloropyridine ≥98.0% (by GC)
3-Amino-2-bromo-5-chloropyridine
Formula: C₅H₄BrClN₂
Peso molecolare: 207,46 g/mol
MDL Number: MFCD07368976
CAS Number: 90902-83-3

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR