Stampa…

6-Bromo-2-chloropyridin-3-amine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Amino-6-bromo-2-chloropyridine

A2967-1G A2967-5G
TCIAA2967-1GEA 20 EUR
TCIAA2967-1G TCIAA2967-5G
6-Bromo-2-chloropyridin-3-amine ≥98.0% (by GC)
6-Bromo-2-chloropyridin-3-amine
Formula: C₅H₄BrClN₂
Peso molecolare: 207,4600 g/mol
MDL Number: MFCD04971930
CAS Number: 169833-70-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR