Stampa…

Ambrisentan ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic acid, (S)-2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid

A2966-20MG A2966-100MG
TCIAA2966-20MGEA 35 EUR
TCIAA2966-20MG TCIAA2966-100MG
Ambrisentan ≥98.0% (by HPLC)
Ambrisentan
Formula: C₂₂H₂₂N₂O₄
Peso molecolare: 378,42 g/mol
MDL Number: MFCD09842330
CAS Number: 177036-94-1
Merck Index: 14,00384

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.00 to 7.80 %
Specific rotation [a]20/D +170 to +176 deg (C=0.5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR