4'-Acetoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Acetic acid 4-Acetyl-3-hydroxyphenyl ester, 4'-Acetoxy-2'-hydroxyacetophenone, 4-Acetyl-3-hydroxyphenyl acetate
A2960-1G
A2960-5G
TCIAA2960-1GEA
57
EUR
TCIAA2960-1G
TCIAA2960-5G
4'-Acetoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
4'-Acetoxy-2'-hydroxyacetophenone
Formula:
C₁₀H₁₀O₄ Punto di ebollizione: 303°C |
MDL Number:
MFCD00092225 CAS Number: 42059-48-3 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...