Stampa…

3-Amino-5-(4-methoxyphenyl)pyrazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2953-1G A2953-5G
TCIAA2953-1GEA 90 EUR
TCIAA2953-1G TCIAA2953-5G
3-Amino-5-(4-methoxyphenyl)pyrazole ≥98.0% (by HPLC, titration analysis)
3-Amino-5-(4-methoxyphenyl)pyrazole
Formula: C₁₀H₁₁N₃O
Peso molecolare: 189,22 g/mol
MDL Number: MFCD03093928
CAS Number: 19541-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6874476bb06193a7a1fd522654d11f1492531ba&vid=b6874476bb06193a7a1fd522654d11f1492531ba&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR