N,N-Dimetilacetammide, anidro ≥99.8%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Packaged under argon in resealable ChemSeal™ bottles.
Purity: 99.8%
CAS No.: 127-19-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 0.940
Refractive Index: 1.4385
Appearance: Colourless
Odour: Ammonia-like
Melting Point: -20°
Boiling Point: 165-166°
Beilstein Reference: 1737614
Merck Reference: 14,3227
MDL No.: MFCD00008686
Formula:
CH₃CON(CH₃)₂ Punto di ebollizione: 165…166 °C Punto di fusione: –20 °C Densità: 0,940 Flash Pt: 70 °C (158 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 UN: 2810 ADR: 6.1,III Merck Index: 14,3227 |