5-Formyl-2-methoxyphenyl Acetate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Acetic acid 5-Formyl-2-methoxyphenyl ester, 3-Acetoxy-4-methoxybenzaldehyde
Formula:
C₁₀H₁₀O₄ Peso molecolare: 194.19 g/mol |
MDL Number:
MFCD00266441 CAS Number: 881-57-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...