Stampa…

4-Chloropyrimidin-5-amine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Amino-4-chloropyrimidine

A2937-200MG A2937-1G
TCIAA2937-200MGEA 36 EUR
TCIAA2937-200MG TCIAA2937-1G
4-Chloropyrimidin-5-amine ≥97.0% (by GC, titration analysis)
4-Chloropyrimidin-5-amine
Formula: C₄H₄ClN₃
Peso molecolare: 129,55 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06412561
CAS Number: 54660-78-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR