Stampa…

3-Amino-6-bromo-2-methylpyridine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

A2928-1G A2928-5G
TCIAA2928-1GEA 28 EUR
TCIAA2928-1G TCIAA2928-5G
3-Amino-6-bromo-2-methylpyridine ≥97.0% (by GC, titration analysis)
3-Amino-6-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
Peso molecolare: 187,04 g/mol
MDL Number: MFCD03095086
CAS Number: 126325-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b43b3a356d005132aec2f08d738081b5ab18071&vid=7b43b3a356d005132aec2f08d738081b5ab18071&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR