3-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Benzylpiperidin-3-amine, 1-Benzylpiperidin-3-ylamine
A2917-1G
A2917-5G
TCIAA2917-1GEA
54
EUR
TCIAA2917-1G
TCIAA2917-5G
3-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
3-Amino-1-benzylpiperidine
Formula:
C₁₂H₁₈N₂ Peso molecolare: 190,29 g/mol Punto di ebollizione: 82°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD04038444 CAS Number: 60407-35-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...