6-Aminomethylquinoline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-(Aminomethyl)quinoline
A2908-200MG
A2908-1G
TCIAA2908-200MGEA
72
EUR
TCIAA2908-200MG
TCIAA2908-1G
6-Aminomethylquinoline ≥98.0% (by GC, titration analysis)
6-Aminomethylquinoline
Formula:
C₁₀H₁₀N₂ Peso molecolare: 158,204 g/mol Punto di ebollizione: 159°C Densità: 1,02 |
MDL Number:
MFCD02853688 CAS Number: 99071-54-2 |
Risultati del test di specificazione
Appearance | Colorless to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...