4-(3,4-Dichlorophenyl)thiazol-2-amine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Amino-4-(3,4-dichlorophenyl)thiazole, 4-(3,4-Dichlorophenyl)-2-thiazolamine
A2906-1G
A2906-5G
TCIAA2906-1GEA
37
EUR
TCIAA2906-1G
TCIAA2906-5G
4-(3,4-Dichlorophenyl)thiazol-2-amine ≥98.0% (by GC, titration analysis)
4-(3,4-Dichlorophenyl)thiazol-2-amine
Formula:
C₉H₆Cl₂N₂S Peso molecolare: 245,1300 g/mol |
MDL Number:
MFCD00466323 CAS Number: 39893-80-6 |
Risultati del test di specificazione
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...